Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Mouse anti-Human, AF 700, Clone: UCHT1, Southern Biotech™
Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 951527
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD3 | |
Monoclonal | |
Lot specific | |
Supplied in PBS/NaN3 | |
Human peripheral blood lymphocytes from a patient with Sézary's syndrome and infant thymocytes | |
RUO | |
Human | |
IgG1 |
Flow Cytometry | |
UCHT1 | |
Alexa Fluor 700 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-8°C; Protect from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction