Learn More
Sino Biological CD28 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 90182C08H1
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
A DNA sequence encoding the human/cynomolgus/rhesus CD28 (P10747-1/Q0PDN3-1/NP_001036106.2) (Asn19-Pro152) was expressed with a polyhistidine tag at the C-terminus and a signal peptide (Q0PDN3-1/NP_001036106.2, Met1-Gly18) at the N-terminus. Human, Cynomolgus and Rhesus CD28 sequences (Asn19-Pro152) are identical.
Catalog No. 50-237-3909