Learn More
Sino Biological CD19 Protein 100ug (Sino Biological)
A DNA sequence encoding the cynomolgus / rhesus CD19 (XP_005591597.1) (Met1-Lys292) was expressed with the Fc region of human IgG1 at the C-terminus. Cynomolgus and Rhesus CD19 sequences are identical.