Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD103 Rat anti-Mouse, BUV395, Clone: M290, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 568715
This item is not returnable.
View return policy
Description
CD103 Monoclonal antibody specifically detects CD103 in Mouse samples. It is validated for Flow cytometry (Routinely Tested)Specifications
CD103 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
16407 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2a κ |
Flow Cytometry | |
M290 | |
Brilliant Ultraviolet 395 | |
Itgae; Integrin alpha-E; αE; alpha-E1; ITAE; Integrin αIEL chain; aM290 | |
Mouse Intestinal Epithelial Cells | |
50 μg | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction