Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ CCND1-FGF19/CEN11p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
$2310.00
Specifications
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
---|---|
Includes | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
For Use With (Application) | FISH-Ce |
Gene ID (Entrez) | 595/9965 |
Format | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-938-486
|
Abnova™
FG0185 |
100 µL |
Each of 1 for $2,310.00
|
|
|||||
Specifications
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
Liquid |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
595/9965 | |
Store at 4°C in the dark. |
Spot an opportunity for improvement?
Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title