Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL21 Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

$211.00 - $270.00

Specifications

Antigen CCL21
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
500P10950UG
Encompass
View Documents
Pepro Tech Inc
500P10950UG
50μg
Each for $211.00
Add to Cart
 
500P109100
Encompass
View Documents
Pepro Tech Inc
500P109100UG
100 μg
Each for $270.00
Add to Cart
 
Description

Description

AA Sequence of recombinant protein: discontinued. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Exodus-2 (CCL21). Anti-Human Exodus-2 (CCL21)-specific antibody was purified by affinity chromatography employing an immobilized Human Exodus-2 (CCL21) matrix. Sandwich ELISA: To detect Human Exodus-2 (CCL21) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Exodus-2 (CCL21) (500-P109Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Exodus-2 (CCL21). Western Blot: To detect Human Exodus-2 (CCL21) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Exodus-2 (CCL21) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils. The cytokine encoded by this gene may also play a role in mediating homing of lymphocytes to secondary lymphoid organs. It is a high affinity functional ligand for chemokine receptor 7 (CCR7) that is expressed on T and B lymphocytes and a known receptor for another member of the cytokine family (small inducible cytokine A19).
Specifications

Specifications

CCL21
ELISA, Western Blot
Unconjugated
Rabbit
Human
O00585
6366
E.coli-derived Recombinant Human Exodus-2 (CCL21)
Antigen affinity chromatography
CCL21
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
6 Ckine; 6CKBAC1; 6CKBAC2; 6Ckine; AI159701; ALP; AW987545; beta chemokine exodus-2; beta-chemokine exodus-2; CC chemokine 6Ckine-ser; C-C motif chemokine 21; C-C motif chemokine 21a; C-C motif chemokine 21b; C-C motif chemokine 21c; C-C motif chemokine ligand 21; CCL21; Ccl21a; Ccl21b; Ccl21c; Ccl21-leu; CCL21-Ser; chemokine (C-C motif) ligand 21; chemokine (C-C motif) ligand 21A (serine); chemokine (C-C motif) ligand 21B (leucine); chemokine (C-C motif) ligand 21b (serine); chemokine (C-C motif) ligand 21C (leucine); chemokine CCL21/6CKINE; CKb9; ECL; Efficient Chemoattractant for Lymphocytes; exodus-2; plt; Scya21; SCYA21a; Scya21b; Scya21c; secondary lymphoid organ chemokine; secondary lymphoid tissue chemokine; Secondary lymphoid-tissue chemokine; SLC; small inducible cytokine A21; small inducible cytokine A21a; small inducible cytokine A21c; small inducible cytokine subfamily A (Cys-Cys), member 21; small-inducible cytokine A21; Small-inducible cytokine A21a; Small-inducible cytokine A21b; Small-inducible cytokine A21c; TCA4; Thymus-derived chemotactic agent 4; UNQ784/PRO1600
CCL21
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.