Learn More
Brady™ Caution Asbestos Hazard Pipe Marker, Style 1HV
Self-sticking Vinyl Pipe Markers meet the requirements of the ASME (ANSI) A13.1 Standard for identifying piping system contents when used with Directional Flow Arrow Tape.
Supplier:  Brady™ 97627PK
Catalog No. 17-200-438
Explore all special offers
Add to Cart
Description
Description
  • Durable B-946 material is ideal for indoor and outdoor environments
  • Style 1HV pipe markers measure 4 x 24 in., come with one marker per card and fit 8 in. or greater pipe diameter
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil and water resistance
Specifications
Specifications
Permanent Acrylic
Black on Yellow
Gloss
Vinyl
Non-sterile
Pipe and Valve Marking
8 to 10 Years
-40°F to +180°F
0.004 in.
101.6 x 609.6 mm
24 in.
3.5 in.
None
Text
Label
Pipe Marker
Caution Asbestos Hazard Do Not Disturb without Proper Training and Equipment
English
1 Marker/Card, 5 Cards/Pk.
-40°C to +82°C
0.102 mm
4 in.
609.6 mm
88.9 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.