Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CASIN

Cdc42 inhibitor

$337.50

Specifications

Purity >98%
Product Type CASIN
Molecular Weight (g/mol) 306.4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-501-0
View Documents
Tocris Bioscience™
5050/10
10 mg
Each for $337.50
Add to Cart
 
50-505-0R
View Documents
Tocris Bioscience™
5050/50
50 mg N/A N/A N/A
Description

Description

Cdc42 GTPase inhibitor (IC50 = 2 μM). Reduces active levels of Cdc42 in aged hematopoietic stem cells (HSCs) to that in young HSCs. Reverses the aging-related and polarity phenotype of aged HSCs to that of young HSCs in vivo. Increases histone H4K16 acetylation in aged HSCs.
Specifications

Specifications

>98%
CASIN
306.4
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.