Learn More
Bioworld Carnivorous Plant (Nepenthes) Basal Salt Mixture, 25 L
Supplier: Bioworld 306300892
Carnivorous Plant (Nepenthes) Basal Salt MixtureContains Micronutrients, Macronutrients, and Vitamins required for Nepenthes growthSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 0.65 g/LSoluble in waterCommonly supplemented with MS Vitamins and Sucrose
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.