Learn More
STEMCELL Technologies Carmine Alum, Size: 100 mL
Supplier: STEMCELL Technologies 07070
Carmine Alum is used for the whole mount staining of mouse mammary gland. This product contains carmine and aluminum potassium sulfate in water. Products are for Research Use Only, Not for Diagnostic or Therapeutic use, and are not to be administered to humans.