Learn More
Enzo Life Sciences Carboxy-PTIO (50mg). CAS: 148819-94-7
Supplier: Enzo Life Sciences ALX430001M050
Water soluble and stable nitric oxide radical scavenger that shows in chemical and biological systems antagonistic action against the free nitric oxide radical (NO·). The inhibitory effects were almost 2-fold stronger than those of NMMA (ALX-106-001) and NOARG (ALX-105-001). Reacts with nitric oxide in a stoichometric manner. Can be used for an improved detection of nitric oxide in activated macrophage cultures in combination with the Griess reagent (ALX-400-004). Protects against endotoxin shock. Alternative name: 2-(4-Carboxyphenyl)-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide . potassium salt. Purity: ≥99% (HPLC). Solubility: Soluble in 100% ethanol, methanol, DMSO or dimethyl formamide. Highly soluble in water or aqueous buffers, e. g. PBS, pH 7.2. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.