Learn More
Enzo Life Sciences Carboplatin (100mg). CAS: 41575-94-4
Supplier: Enzo Life Sciences ALX400041M100
An analog of cisplatin (ALX-400-040). Antitumor agent. Alternative name: cis-Diammine [1,1-cyclobutanedicarboxylato] platinum (II). Purity: ≥98% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.