Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XIV/CA14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | Carbonic Anhydrase XIV/CA14 |
---|---|
Dilution | Western Blot 1:500-1:1000, ELISA 1:5000-1:10000 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Carbonic Anhydrase XIV/CA14 Polyclonal antibody specifically detects Carbonic Anhydrase XIV/CA14 in Mouse samples. It is validated for Western Blot, ELISASpecifications
Carbonic Anhydrase XIV/CA14 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
0.2 um filtered solution in PBS | |
23632 | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:500-1:1000, ELISA 1:5000-1:10000 | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1 | |
Produced in rabbits immunized with purified, recombinant Mouse Carbonic Anhydrase XIV/CA14 (Accession#: NP_035927.1; Met 1-Met 290) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title