Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VII/CA7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | Carbonic Anhydrase VII/CA7 |
---|---|
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Carbonic Anhydrase VII/CA7 Polyclonal antibody specifically detects Carbonic Anhydrase VII/CA7 in Human samples. It is validated for ELISASpecifications
Carbonic Anhydrase VII/CA7 | |
ELISA | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
0.2 um filtered solution in PBS | |
766 | |
IgG | |
Antigen and protein A Affinity-purified |
ELISA 1:5000-1:10000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Carbonate dehydratase VII, carbonic anhydrase 7, carbonic anhydrase VIICAVII, carbonic dehydratase VII, CA-VII, EC 4.2.1.1 | |
Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase VII/CA7 (Accession#: P43166; Met1-Ala264) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title