Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase III Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody

$182.80 - $408.00

Specifications

Antigen Carbonic Anhydrase III
Clone C2
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FEMA550993
Encompass_Preferred
View Documents
Thermo Scientific
MA550993
200 μL
Each for $408.00
Add to Cart
 
FEMA550994
Encompass_Preferred
View Documents
Thermo Scientific
MA550994
20 μL
Each for $182.80
Add to Cart
 
Description

Description

Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10. 3 kb and contains seven exons and six introns.
Specifications

Specifications

Carbonic Anhydrase III
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4
BB219044, CA III, Ca3, CAH3, CAIII, CA-III, CAIII (Muscle), Car3, Car-3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, carbonic anhydrase-like protein, EC 4.2.1.1, epididymis secretory sperm binding protein Li 167mP, HEL-S-167mP
Ca3, Car3
IgG2b κ
Protein A/G
Ca3
C2
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse, Porcine, Rat
P07451, P14141, P16015, Q5S1S4
12350, 494016, 54232, 761
Recombinant Human CA3 protein, Ala2-Lys260 (Accession #P07451)
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.