Learn More
Bioworld Carbonate-buffered saline pH 9.5 (5X), 500 mL
Supplier: Bioworld 403200272
This buffer contains 125mM sodium carbonate and 750mM sodium chloride, pH 9.5. The buffer is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it.Warning:For laboratory use only.Not fit for agricultural, household, clinical use or humanconsumption. Appearance: LiquidPH Value: 9.5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.