Learn More
Sklar Instruments Disposable Carbon Steel Surgical Blades Sklar 15. Sklar Disposable, Sterile, Carbon Steel, Box of 100.
Supplier: Sklar Instruments 063015
Disposable Carbon Steel Surgical Blades Sklar, #15. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #15 is most commonly used for short or curved incisions. Disposable blades are available in bulk quantities of 100 per box.