Learn More
OHAUS Carbon Steel Defender 5000 Series Floor Scale
Defender 5000 Series Floor Scales meet advanced weighing needs for industrial and commercial use. Their high-quality construction is designed to minimize flexing under heavy loads for years of dependable performance. The indicator features software for counting, checkweighing, accumulation, display hold, and other modes.
Supplier: OHAUS 30837699
Description
- Painted carbon steel, treaded decks with four alloy steel load cells, fixed top plate (5 mm/0.196 in. thick) with structural bracing for stiff construction.
- Platforms available in sizes of 3 x 3 ft. and 4 x 4 ft. and capacities from 2,500 lb. to 5,000 lb. with NTEP certified 5,000e performance, allowing greater flexibility for your weighing needs.
- Equipped with a new TD52 series indicator featuring an easy-to-use keypad, large backlit display, and plain text on-screen prompts to guide you through operation.
Specifications
2500 kg | |
0.5 kg | |
Scale |
LCD | |
i-DF52P5000B1L | |
Class III |