Learn More
World Precision Instrument Carbon, 18 AWG, red
Supplier: World Precision Instrument CPVC4050R
Carbon filament wire is made of 3,000 microscopic carbon strands in a red PVC jacket. 45.8 ohms/foot.