Learn More
Volu Sol Carbol Xylene, 16 oz / 500 mL, Volu-Sol™
Supplier: Volu Sol VXZ016
Used for intestinal protozoa determination. Component of Volu-Sol's Parasitology Kit. Has many uses in histology and cytology