Learn More
Pfanstiehl Inc D-Mannitol, (Plant Derived) High Purity / Low Endotoxin-Low Metals USP/EP/JP/ChP (M-109-7) - 5kg pack GMP, Multicompendial CAS: 69-65-8
Supplier:  Pfanstiehl Inc M10975KG
Encompass_Preferred
Mannitol is classified as a sugar alcohol and is derived from a sugar (mannose) by reduction. The physical properties of mannitol make it suitable for use as a bulking agent in tableting. It also has unique chemical properties that enable it to stabilize a wide variety of pharmaceutical preparations, including liquid formulations. It has been utilized as a key component to stabilize proteins during formulation, freeze drying, and fill/finish applications.Pfanstiehl High Purity Low Endotoxin Mannitol was developed specifically for biopharmaceutical applications, to the highest quality and purity standards. The various benefits imparted of Mannitol include but are not limited to: Enhanced Protein Therapeutic Stability Vaccine Stabilization Carrier in Lyophilized & Spray Dried FormulationsPfanstiehl Mannitol is available as NF (National Formulary), EP (European Pharmacopoeia), JP (Japanese Pharmacopoeia), and ChP (Chinese Pharmacopoeia).
Catalog No. 50-165-6645
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.