Learn More
Andwin Scientific CAPS-PISTONS 50-250 MICROL. 3C
SDP
Supplier:  Andwin Scientific C250
Encompass_Preferred
The Positive Displacement tips are presterilized and preassembled. The tips have a maximum volume capacity of 250 L and have capillaries with pistons tip technology. This product is specially designed to be compatible with Pos-D and Gilson Microman positive-displacement pipettes.
Catalog No. NC0128894
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.