Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPRIN1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$150.28
Specifications
Antigen | CAPRIN1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-6310
|
Bethyl Laboratories, Inc
A303881AT |
20 μL |
Each of 1 for $150.28
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.Specifications
CAPRIN1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Caprin-1; Cytoplasmic activation- and proliferation-associated protein 1; GPI-anchored membrane protein 1; GPI-anchored protein p137; p137GPI; Membrane component chromosome 11 surface marker 1; CAPRIN1; GPIAP1; GPIP137; M11S1 | |
Between 450 and 500 | |
Primary | |
4° C | |
CAPRIN1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q14444, Q60865 | |
4076, 53872 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title