Learn More
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kit
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kits enable convenient collection and transport of samples for drugs of abuse or forensic testing.
Supplier: Thermo Scientific™ 30104
Description
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kits enable convenient collection and transport of samples for drugs of abuse or forensic testing. Each kit contains a collection foil, transport envelope, specimen bag, and tamper-evident tape.
Features:
- Tamper-evident closures
- Collection containers are Class 10,000 aseptic products
- Customized kits and OEM parts available
- Leak-resistant & air-tight, with a lock seal
- Twin valve design provides tighter seal
Specifications
Absorbent 3 in. x 6 in. Cut |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.