Learn More
Boston Bioproducts Inc Calcium Chloride (1 M) - 100ml
Supplier: Boston Bioproducts Inc MT140100ML
The 1M Calcium Chloride solution allows for the adjustment of calcium required by cultured cells, and is effective in creating environments that facilitate ionic strength and osmotic pressure. Calcium chloride is commonly used as a source of calcium ions in the preparation and transformation of competent cells, the formulation of cell culture media, in enzymatic reactions, and protoplast preparation. Its unique properties also make it suitable for use in many molecular biology applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.