Learn More
Enzo Life Sciences Cabazitaxel (100mg). CAS: 183133-96-2
SDP
Supplier:  Enzo Life Sciences ENZCHM2170100
Encompass_Preferred
Cabazitaxel is a semi-synthetic derivative of a natural taxoid Purity: ≥99% (HPLC). Solubility: Soluble in DMSO at 100 mg/mL. Long Term Storage: -20°C.
Catalog No. 50-201-3559
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.