Learn More
Enzo Life Sciences Complement component C5b-9 monoclonal antibody (aE11) (biotin conjugate) (150 µg)
SDP
Supplier:  Enzo Life Sciences BPDDIA01101B015
Encompass_Preferred
Synonyms: Terminal complement complex, TCC. Clone: aE11. Host: Mouse. Isotype: IgG2aκ. Immunogen: Purified human complement component C5b-9. Species reactivity: Human, Porcine, Horse, Baboon. Applications: ELISA. Purification: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.14M sodium chloride and 15mM sodium azide.
Catalog No. 50-210-0402
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.