Learn More
As One International Inc C57BL/6 Mouse Mesenchymal Stem Cells with RFP, 1×10^6/vial
Supplier:  As One International Inc MUBMX01201
Encompass_Preferred
Mesenchymal Stem Cells (MSCs) are multipotent stem cells that can differentiate into a variety of cell types, including osteocytes, adipocytes, and chondrocytes. MSCs porliferate quickly and are capable of generating a local immunosuppressive microenvironment, which contributes to their wide application potentials in tissue engineering, cell therapy and gene therapy.
Catalog No. 50-165-5012
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.