Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C17orf62 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA561073
Description
Immunogen sequence: MVVLRLATGF SHPLTQSAVM GHRSDVEAIA KLITSFLELH CLESPTELSQ SSDSEAGDPA SQS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
Specifications
C17orf62 | |
Polyclonal | |
Unconjugated | |
C17ORF62 | |
C17orf62; chromosome 17 open reading frame 62; CYBC1; Cytochrome b-245 chaperone 1; EROS; Essential for reactive oxygen species protein; HAPSTR1; uncharacterized protein C17orf62 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79415 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BQA9 | |
C17ORF62 | |
Recombinant protein corresponding to Human C17orf62 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction