Learn More
Enzo Life Sciences C. difficile toxin A (50µg)
SDP
Supplier:  Enzo Life Sciences BMLG1400050
Encompass_Preferred
Potent enterotoxin. Potent enterotoxin. Alternative name: Clostridium difficile toxin A. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains 0.05M HEPES, 0.15M sodium chloride and 5% sucrose. Source: From Clostridium difficile, strain VPI10463. Long Term Storage: +4°C. Applications: ELISA LINE assay, Cytotoxicity studies, Antibody capture.
Catalog No. 50-201-0845
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.