Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV605 Rat Anti-Mouse TNF, BD Horizon™

Rat Monoclonal Antibody

$152.00 - $252.00

Specifications

Antigen TNF
Clone MP6-XT22
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB569296
View Documents
BD Biosciences
569296
0.1 mg
Each for $252.00
Add to Cart
 
BDB569297
View Documents
BD Biosciences
569297
25 μg
Each for $152.00
Add to Cart
 
Description

Description

TNF Monoclonal antibody specifically detects TNF in Mouse samples. It is validated for Intracellular staining (flow cytometry)
Specifications

Specifications

TNF
0.2 mg/mL
Monoclonal
Liquid
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
21926
IgG1
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
MP6-XT22
Flow Cytometry
Brilliant Violet 605
Rat
Mouse
Tnf; Tnfa; TNF alpha; TNF-a; Tnfsf1a; Tnfsf2; TNFSF2; Cachectin; DIF
Recombinant Mouse TNF
Affinity Chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.