Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV510 Mouse Anti-Human CD34, BD Horizon™
Mouse Monoclonal Antibody
$168.00 - $412.00
Specifications
Antigen | CD34 |
---|---|
Clone | QBEND/10.rMAb (also known as QB-END-10, QBEND10, QBEND/10, QBEnd10) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 510 |
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Flow cytometrySpecifications
CD34 | |
Flow Cytometry | |
Brilliant Violet 510 | |
Mouse | |
Human | |
CD34; CD34 antigen | |
Human Placental Vesicles | |
Affinity Chromatography |
QBEND/10.rMAb (also known as QB-END-10, QBEND10, QBEND/10, QBEnd10) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
947 | |
IgG1 λ2 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title