Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Rat Anti-Mouse CD8b, BD Horizon™

Rat Monoclonal Antibody

$172.00 - $426.00

Specifications

Antigen CD8b
Clone YTS156.7.7.rMAb (also known as YTS156.7.7; YTS156)
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB569871
View Documents
BD Biosciences
569871
0.1 mg
Each for $426.00
Add to Cart
 
BDB569947
View Documents
BD Biosciences
569947
25 μg
Each for $172.00
Add to Cart
 
Description

Description

CD8b Monoclonal antibody specifically detects CD8b in Mouse samples. It is validated for Flow cytometry
Specifications

Specifications

CD8b
0.2 mg/mL
Monoclonal
Liquid
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
12526
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
YTS156.7.7.rMAb (also known as YTS156.7.7; YTS156)
Flow Cytometry
Brilliant Violet 421
Rat
Mouse
Cd8b1; Cd8b; Ly-3; Lyt-3; lymphocyte antigen 3
Mouse Thymocytes
Affinity Chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.