Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Mouse Anti-Human TIGIT, BD Horizon™

Mouse Monoclonal Antibody

$232.00 - $486.00

Specifications

Antigen TIGIT
Clone TgMab-2
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Violet 421
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB570375
View Documents
BD Biosciences
570375
100 Tests
Each for $486.00
Add to Cart
 
BDB570445
View Documents
BD Biosciences
570445
25 Tests
Each for $232.00
Add to Cart
 
Description

Description

TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow cytometry
Specifications

Specifications

TIGIT
Flow Cytometry
Brilliant Violet 421
Mouse
Human
T-cell immunoreceptor with Ig and ITIM domains; VSIG9; VSTM3; WUCAM
Human TIGIT Transfected Cell Line
Affinity Chromatography
TgMab-2
Monoclonal
Liquid
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
201633
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.