Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUV395 Mouse Anti-Human CD27, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569712
Description
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow cytometrySpecifications
CD27 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
M-T271 | |
Brilliant Ultraviolet 395 | |
TNFRSF7; TNF receptor superfamily, member 7; T14; Tp55; S152 | |
Human T-CLL cells | |
50 μg | |
939 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction