Learn More
Chemglass Life Sciences Bushing, #50, Internal, Front Seal, Solid, PTFE, #225 Viton O-Ring
SDP
Supplier:  Chemglass Life Sciences CG36614
Encompass_Preferred
Bushing, #50, Internal, Front Seal, Solid, PTFE, #225 Viton O-Ring. New front seal PTFE bushing for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings listed under CG-362.
Catalog No. 50-873-547
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.