Learn More
Med Vet International Miltex Friction Grip Diamond Bur, Flat End Cylinder, Medium Grit, German, Each, 835/008
Supplier: Med Vet International MIL835/008
Miltex diamond burs are produced using a coating technique, which assures approximately 85% of the working surface is coated with a uniform diamond grit. Flat end cylinder.