Learn More
Hach Company Citrate Buffer Powder Pillows for Manganese, 25 mL, pk/100
Supplier:  Hach Company 98399
Encompass_Preferred
For determination of manganese by the periodate oxidation method. EPA approved for reporting wastewater analyses (digestion required). Hach method 8034. Sample size: 25 mL Pack of 100 Powder Pillows.
Catalog No. 50-199-4485
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.