Learn More
Accuflow Systems BTL CARRIER RBR 1 GAL R
Supplier:  Accuflow Systems B100 RED
Encompass_Preferred
THIS BOTTLE CARRIER IN RED ACCOMMODATES 2.5 LITER (5 PINT) AND 4 LITER (1 GALLON) BOTTLES AND RESISTS MOST AQUEOUS SOLUTIONS, ACIDS AND BASES. MADE FROM A UNIQUE, HIGH PERFORMANCE THERMOPLASTIC ELASTOMER, IT HAS EXCELLENT TENSILE STRENGTH AND CAN BE USED FROM -45C TO +110C. IT IS ALSO STEAM STERILIZABLE, RESISTS OXIDATION IN PROLONGED ATMOSPHERIC EXPOSURE AND HAS A HANDLE MOLDED AS PART OF THE CARRIER. A RECESSED MARKING AREA AT THE TOP OUTER SIDE OF THE CARRIER WHICH CAN BE USED TO IDENTIFY THE LIQUID BEING CARRIED OR ICE WITHOUT SWEATING. IT RESISTS BACTERIAL AND FUNGI GROWTH. THE HANDLE IS PRECISION-MOLDED TO ASSURE UNIFORM APPEARANCE AND QUALITY AND EACH CARRIER IS INDIVIDUALLY PACKAGED. A CENTERING RING IN THE BASE HOLDS THE BOTTLE TO ASSURE CLEARANCE FROM THE WALL.
Catalog No. NC1266541
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.