Learn More
New England Biolabs, Inc. BstNI – 15000 units
SDP
Supplier:  New England Biolabs, Inc. R0168L
Encompass
A restriction endonuclease that recognizes the sequence CC^W_GG.
Catalog No. 50-812-074
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.