Learn More
New England Biolabs, Inc. Bst 2.0 WarmStart DNA Polymerase (Glycerol-free) - 8000 units
SDP
Supplier:  New England Biolabs, Inc. M0402L
Encompass
Bst 2.0 WarmStart DNA Polymerase (Glycerol-free) is formulated without glycerol to support lyophilization, incorporation into microfluidic devices, and enable quick adoption into automation workflows. Bst 2.0 WarmStart DNA Polymerase is an in silico designed homologue of Bacillus stearothermophilus DNA Polymerase I, Large Fragment (Bst DNA Polymerase, Large Fragment) with a reversibly-bound aptamer, which inhibits polymerase activity at temperatures below 45°C. At temperatures above 45°C, the aptamer rapidly releases from the enzyme with no special activation step needed to activate the polymerase. Bst 2.0 WarmStart DNA Polymerase contains 5' to 3' DNA polymerase activity and strong strand-displacement activity but lacks 5' to 3' exonuclease activity. Bst 2.0 WarmStart DNA Polymerase displays improved amplification speed, yield, salt tolerance, and thermostability compared to wild-type Bst DNA Polymerase, Large Fragment.
Catalog No. 50-258-1812
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.