Learn More
Andwin Scientific BRUSH 5/PK
SDP
Supplier:  Andwin Scientific 17150041
Encompass_Preferred
The brushes by Mill-Rose provide thorough cleaning of bottles and flasks. The ends of the natural bristle brush head have radial tips, which permit users to scrub the area of the desired labware more efficiently. The black bristles are securely attached into galvanized wire. The brush has a diameter of 4.4 cm (13/4") and has an overall length of 41 cm (16"). This is sold in a pack of 5.
Catalog No. 50-101-105
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.