Learn More
Research Products International Corp Brembox, 3/8 Inch Thick, Capacity 30 1.5 ml Tubes
Supplier: Research Products International Corp BR009
This stackable container provides shielding from beta radiation and allows efficient use of freezer space. Removable cover also serves as a portable shield. Holds 1.5 ml - 2.0 ml micro-tubes. Safe for freezer storage to -70?C.