Learn More
Electron Microscopy Sciences Brain Punch Set of Three, 1.50 to 2.00 mm
Supplier: Electron Microscopy Sciences 57402
The Brain Punch Kit is a set of electropolished and sharpened punches for the Palkovit's punch technique. Complete with spring loaded nylon expellers and a convenient handle. The nuclei are punched from frozen brain sections on glass slides, and expelled into the reagents. The spring-loaded expellers avoid risk of contamination that might be caused by expelling with breath.
Size: 1.50 to 2.00 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.