Learn More
Electron Microscopy Sciences Brain Punch Individual, 1.25 mm
Supplier:  Electron Microscopy Sciences 57399
Encompass_Preferred
Electropolished and sharpened punches for the Palkovit's punch technique.
Size: 1.25 mm
Catalog No. 50-192-6844
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.