Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Brace-Bow™ Stand-Up Floor Sign - COVID-19 VACCINATIONS (English, Spanish), Plastic

Brace-Bow™ stand-up floor signs communicate health and safety policies, reinforce procedures and identify locations

Supplier:  Accuform™ SBPFL234

 View more versions of this product

Catalog No. 19-119-201


Description

Description

  • Upright, stand-up sign has bold message printed on one side
  • High-impact, thick plastic panels adhere together at top
  • Sturdy panels bow and flex towards bottom
  • 14 in. bottom fastens together with hook and loop attachment at base
  • For transport and flat storage, remove base and close panels
  • Weighing nearly 10 lb., sign heavy enough to withstand light wind conditions
  • Specifications

    Specifications

    Plastic
    Floor-standing
    24 in.
    English, Spanish
    Blue on White
    Left arrow
    36 in.
    Rectangular
    COVID-19 VACCINATIONS (English, Spanish)
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Promotions

    Promotions

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.