Learn More
Enzo Life Sciences ET A receptor antagonist [BQ-123]. sodium salt (5mg). CAS: 136553-81-6
SDP
Supplier:  Enzo Life Sciences ALX155004P005
Encompass_Preferred
Highly potent and selective ETA antagonist. Formula: C31H42N6O7 . Na. MW: 633.7. Purity: ≥96%. Solubility: Soluble in water. Long Term Storage: -20°C. Use/Stability: Stable for at least 3 years after receipt when stored at -20°C. Stock solutions are stable for up to 3 months when stored at -20°C.
Catalog No. 50-200-7797
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.