Learn More
Electron Microscopy Sciences Bouin's Solution 4 LT
Supplier: Electron Microscopy Sciences 1599004
Bouin's solution can be used as a fixation and a staining fluid and is excellent for use on biopsy specimens of the gastrointestinal tract. Tissue from the endocrine system are well fixed and many antibodies react well with tissue fixed in Bouin's. May be used as a routine fixative or a staining solution, but should not be used as a preservative for tissue. The maximum fixation time in this solution should be less than 48 hours and should be transferred to alcohol. Packaged in leak-proof histo-containers, prefilled and ready-to-use.
Formulated with Picric Acid saturated aqueous solution 75%, Formalin (40% aqueous formaldehyde) 25% and Glacial Acetic acid 5% in de-ionized water.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.