Learn More
Electron Microscopy Sciences Boron Carbide Powder, 42 µm particles 1 LB
Supplier:  Electron Microscopy Sciences 5051042
Encompass_Preferred
Boron Carbide is one of the hardest materials known, ranking third behind diamond and cubic boron nitride. This makes it is ideal for use as an abrasive in polishing and lapping applications. Extreme hardness (Knoop 100g: 2900-3580 kg/mm2), Good chemical resistance, Good nuclear properties, Low density (2.52 g/cm2)
Particles: 42 µm
Catalog No. 50-997-434
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.