Learn More
Chemglass Life Sciences BNC Female to Male Extension Cable, 4ft
Supplier: Chemglass Life Sciences CG1872A20
BNC Female to Male Extension Cable, 4ftBNC extension cable for CG-1872-A series of electrode adapters. Adapter permits CG-1872 series pH electrode(12mm OD) to be inserted either directly into smaller reactors or down through a CG-1872-B baffle.